Magnesium chloride 1 mol/l (2 N) in aqueous solution
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:7786-30-3
:
Formula:MgCl₂
MDL Number:MFCD00011106
:95.21 g/mol
Merck Index:13,05684
Storage Temperature:Ambient
EINECS:232-094-6
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together