Butyl hexanoate
Catalog # EHERC10948031
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
Boiling Pt:207 °C (1013 hPa)
Density:0.866 g/cm³ (25 °C)
:172.27 g/mol
EINECS:210-964-6
CAS Number:626-82-4
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00053804
Boiling Pt:207 °C (1013 hPa)