Copper standard solution, 1000 mg/L Cu in dil. nitric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Troclosene sodium, (max. 3% H₂O) 97% (dry weight)
Supplier: Thermo Fisher Scientific