Potassium standard solution, 10,000 mg/L K in dilute nitric acid standard for ICP
: CPACHEM
Frequently Bought Together
pH buffers, ready to use
: WTW
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Picric acid 10 g/l in aqueous solution Reag. Ph. Eur. 1065801
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
Conductivity standards
: HAMILTON BONADUZ
VWR®, Turbidity standards, formazin
: VWR Chemicals
Electrolyte solutions
: Hach
Buffer solutions in sachets, perpHect® Pouches, Orion™
: Thermo Orion
Magnesium ICP standards, (CRMs), Fluka™
: Honeywell Chemicals