Rhodium standard solution, 10,000 mg/l Rh in 10% hydrochloric acid standard for ICP
Supplier: CPACHEM
Certificates
Rhodium standard solution, 10,000 mg/l Rh in 10% hydrochloric acid standard for ICP
Supplier: CPACHEM
About this item
Frequently Bought Together
VWR®, Refractive index Certified Reference Material at 20, 25 and 30 °C
Supplier: VWR Chemicals
Buffer solutions
Supplier: Hanna
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
Sulphaquinoxaline
Supplier: EHRENSTORFER
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
VWR®, Sulphur, Certified Reference Materials, in light mineral oil
Supplier: VWR Chemicals
Buffer PH 4,00 1 * 500 mL
Supplier: REAGECON
Supelco® Organic Reference Standard, α-Tocotrienol
Supplier: Merck
Tefuryltrione 1 * 50 mg
Supplier: EHRENSTORFER
Perfluoropropanesulfonic Acid Sodium
Supplier: EHRENSTORFER
Tridemorph
Supplier: EHRENSTORFER