Order Entry
Austria
ContactUsLinkComponent
Silver standard solution, 1000 mg/L Ag in dil. nitric acid standard for AAS and ICP
Silver standard solution, 1000 mg/L Ag in dil. nitric acid standard for AAS and ICP
Catalog # CPIIS4400-1000511
Supplier:  CPI
CAS Number:  
Silver standard solution, 1000 mg/L Ag in dil. nitric acid standard for AAS and ICP
Catalog # CPIIS4400-1000511
Supplier:  CPI
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Silver
  • Symbol:
    Ag

Specifications

About this item