Order Entry
Austria
ContactUsLinkComponent
 
Di-n-octyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D)
  PROOCILDLM16301.2
 :  
Di-n-octyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D)
  PROOCILDLM16301.2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    93952-13-7
  • Description:
    Di-n-octyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D)
  • Formula:
    C₂₄H₃₈O₄
  • Description:
    Di-n-octyl phthalate-[ring-D4]