Order Entry
Austria
Orders LinkContactUsLinkComponent
Methyl-3,3-dimethoxypropionate

Methyl-3,3-dimethoxypropionate

Glass bottle for solids, 25 g
Catalog # MOLE10262793-25G
Marketsource
Supplier:   Molekula
Methyl-3,3-dimethoxypropionate
Catalog # MOLE10262793-25G
Supplier:  Molekula
Marketsource
CAS Number:  7424-91-1
Methyl-3,3-dimethoxypropionate

Glass bottle for solids, 25 g

CAS Number:  7424-91-1
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Methyl 3,3-dimethoxypropanoate, Methyl 3,3-dimethoxypropionate
  • CAS Number:7424-91-1
  • Formula:C₆H₁₂O₄
  • MDL Number:MFCD00010650
  • Molecular Weight:148.16 g/mol
  • EINECS:231-055-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.