Order Entry
Austria
ContactUsLinkComponent
 
Strontium standard solution, 100 mg/L Sr in dil. nitric acid standard for ICP-MS
Catalog # ACSDM356.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Strontium standard solution, 100 mg/L Sr in dil. nitric acid standard for ICP-MS
Catalog # ACSDM356.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Strontium
  • Symbol:
    Sr

Specifications

About this item