Order Entry
Austria
ContactUsLinkComponent
 
Lanthanum standard solution, 100 mg/L La in dil. nitric acid standard for ICP-MS
  89168.180
 :  CPACHEM
 :  
Lanthanum standard solution, 100 mg/L La in dil. nitric acid standard for ICP-MS
  89168.180
 :  CPACHEM
 :  

 

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Lanthanum
  • Symbol:
    La