Order Entry
Austria
ContactUsLinkComponent
Copper standard solution, 100 mg/L Cu in dil. nitric acid standard for ICP-MS
Copper standard solution, 100 mg/L Cu in dil. nitric acid standard for ICP-MS
Catalog # ACSDM315.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Copper standard solution, 100 mg/L Cu in dil. nitric acid standard for ICP-MS
Catalog # ACSDM315.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Copper
  • Symbol:
    Cu

Specifications

About this item