Order Entry
Austria
ContactUsLinkComponent
 
Tantalum standard solution, 100 mg/L Ta, standard for ICP-MS
Catalog # ACSDM357.2N05FP.L1
Supplier:  CPACHEM
CAS Number:  
Tantalum standard solution, 100 mg/L Ta, standard for ICP-MS
Catalog # ACSDM357.2N05FP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃/0,5% HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Tantalum
  • Symbol:
    Ta

Specifications

About this item