Palladium standard solution, 10,000 mg/L Pd in 5% hydrochloric acid standard for ICP
: CPACHEM
Frequently Bought Together
Thiamphenicol certified 1 * 0,1 g
: EHRENSTORFER
CEFAZOLIN SODIUM SALT 1 * 75 mg
: Merck
Dienochlor certified 1 * 0,25 g
: EHRENSTORFER
in Acetonitrile 1 * 1 mL
: EHRENSTORFER
Pymetrozine certified 1 * 0,1 g
: EHRENSTORFER
Nitenpyram Analytical standard, reference material, Fluka™
: Honeywell Chemicals
Cadusafos
: EHRENSTORFER
Perfluoropropanesulfonic Acid Sodium
: EHRENSTORFER