Order Entry
Austria
ContactUsLinkComponent
Scandium standard solution, 100 mg/L Sc in dil. nitric acid standard for ICP-MS
Scandium standard solution, 100 mg/L Sc in dil. nitric acid standard for ICP-MS
Catalog # ACSDM351.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Scandium standard solution, 100 mg/L Sc in dil. nitric acid standard for ICP-MS
Catalog # ACSDM351.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Scandium
  • Symbol:
    Sc

Specifications

About this item