Order Entry
Austria
ContactUsLinkComponent
 
Manganese standard solution, 100 mg/L Mn in dil. nitric acid standard for ICP-MS
  ACSDM333.2NP.L1
 :  CPACHEM
 :  
Manganese standard solution, 100 mg/L Mn in dil. nitric acid standard for ICP-MS
  ACSDM333.2NP.L1
 :  CPACHEM
 :  

 

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Manganese
  • Symbol:
    Mn