Titanium trialuminide ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Titanium aluminide
CAS Number:12004-78-3
:
Formula:Al₃Ti
MDL Number:MFCD00054094
Storage Temperature:Ambient
EINECS:234-461-6
UN:3178
ADR:4.1,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together