Order Entry
Austria
ContactUsLinkComponent
 
Methyl-4-formylbenzoate for synthesis, Sigma-Aldrich®
Catalog # 8.18490.0100
Supplier:  Merck
CAS Number:  
Methyl-4-formylbenzoate for synthesis, Sigma-Aldrich®
Catalog # 8.18490.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    56 - 59 °C
  • Molar Mass:
    164.16 g/mol
  • Bulk Density:
    528 kg/m3
  • Boiling Point:
    264 - 266 °C (1013 hPa)
  • Flash Point:
    128 °C
  • Density:
    1.2 g/cm3 (20 °C)
  • Ignition Point:
    240 °C

Specification Test Results

About this item

 :
 :164.16 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:216-385-5
CAS Number:1571-08-0
Formula:C₉H₈O₃
MDL Number:MFCD00006950