
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17096-07-0
:
Formula:C₁₆H₃₈O₅Si₄
MDL Number:MFCD00053871
:422.82 g/mol
Boiling Pt:112…115 °C (0.2 mmHg)
Density:0.930
Flash Pt:201 °C (393 °F)
EINECS:241-165-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together