1-Octylmagnesium chloride 2 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:38841-98-4
:
Formula:C₈H₁₇ClMg
MDL Number:MFCD00000476
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:254-147-2
UN:2924
Frequently Bought Together