Ethyl isocyanoacetate 97%
:2999-46-4
:
:C₅H₇NO₂
:MFCD00000007
:113.12 g/mol
:194…196 °C (760 mmHg)
:1.03 g/cm³
:84 °C
:Refrigerator
:221-077-9
:2810
:6.1,III
:GHS07
- Environmentally Preferable:
- Appearance:Clear amber to dark brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4170 to 1.4250 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together