4-Acetamidothiophenol, Sigma-Aldrich®
Catalog # SIALA8201-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.2282 g/mol
:M-Clarity™ quality level = MQ100
EINECS:214-427-7
CAS Number:1126-81-4
Formula:C₈H₉NOS
MDL Number:MFCD00004848