Order Entry
Austria
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Dimethachlor
Supelco® PESTANAL® Organic Reference Standard, Dimethachlor
Catalog # 45447-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Dimethachlor
Catalog # 45447-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    50563-36-5
  • Description:
    Dimethachlor
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H317 - H330 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055472
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    255,74
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P260 - P273 - P280 - P301 + P312 - P302 + P352 - P304 + P340 + P310