
3-Cyano-1-propylboronic acid pinacol ester 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:238088-16-9
:
Formula:C₁₀H₁₈BNO₂
MDL Number:MFCD09953480
:195.07 g/mol
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS06
Related Information
Frequently Bought Together