Phenyl glycidyl ether, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Glycidyl phenyl ether
CAS Number:122-60-1
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00005133
Molecular Weight:150.18 g/mol
:M-Clarity™ quality level = MQ200
EINECS:204-557-2
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together