Order Entry
Austria
ContactUsLinkComponent
Boron standard solution, 1,000 mg/L B in dil. nitric acid standard for ICP
Boron standard solution, 1,000 mg/L B in dil. nitric acid standard for ICP
Catalog # ACSDC005.2NP.L25
Supplier:  CPACHEM
CAS Number:  
Boron standard solution, 1,000 mg/L B in dil. nitric acid standard for ICP
Catalog # ACSDC005.2NP.L25
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item