Strontium standard solution, 1000 mg/L Sr in water standard for ion chromatography
: CPACHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
: HAMILTON BONADUZ
pH buffer solutions
: Mettler - Toledo
VWR®, Primary colour solutions, Reagent Ph. Eur.
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
VWR®, Conductivity standard solutions
: VWR Chemicals