

1,2-Epoxycyclopentane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:285-67-6
:
Formula:C₅H₈O
MDL Number:MFCD00005161
:84.12 g/mol
Boiling Pt:102 °C (760 mmHg)
Density:0.964 g/cm³
Flash Pt:10 °C
Storage Temperature:Refrigerator
EINECS:206-005-6
UN:1993
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4330 to 1.4360 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together