Order Entry
Austria
ContactUsLinkComponent
3,5-Dimethyl-1H-pyrazole-1-methanol, Sigma-Aldrich®
3,5-Dimethyl-1H-pyrazole-1-methanol, Sigma-Aldrich®
Catalog # SIAL331457-25G
Supplier:  Merck
CAS Number:  
3,5-Dimethyl-1H-pyrazole-1-methanol, Sigma-Aldrich®
Catalog # SIAL331457-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3,5-Dimethyl-1-(hydroxymethyl)-1H-pyrazole
 :
 :M-Clarity™ quality level = MQ100
EINECS:286-553-0
CAS Number:85264-33-1
Formula:C₆H₁₀N₂O
MDL Number:MFCD00020738