
1H-Pyrazole-4-carboxylic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37718-11-9
:
Formula:C₄H₄N₂O₂
MDL Number:MFCD00011558
:112.09 g/mol
Melting Pt:281…283 °C
EINECS:253-641-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together