Ethyl-3-amino-1H-pyrazole-4-carboxylate, Sigma-Aldrich®
Catalog # SIALA45009-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate
:
:155.16 g/mol
:M-Clarity™ quality level = MQ100
EINECS:230-262-3
CAS Number:6994-25-8
Formula:C₆H₉N₃O₂
MDL Number:MFCD00005238