Order Entry
Austria
ContactUsLinkComponent
 
Methyl-3-aminopyrazine-2-carboxylate, Sigma-Aldrich®
Catalog # SIAL276154-10G
Supplier:  Merck
CAS Number:  
Methyl-3-aminopyrazine-2-carboxylate, Sigma-Aldrich®
Catalog # SIAL276154-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Methyl 2-aminopyrazine-3-carboxylate
 :
 :153.14 g/mol
EINECS:240-387-5
CAS Number:16298-03-6
Formula:C₆H₇N₃O₂
MDL Number:MFCD00010097
 :M-Clarity™ quality level = MQ100