L-Glutamic acid 5-benzyl ester 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1676-73-9
:
Formula:C₁₂H₁₅NO₄
MDL Number:MFCD00002633
:237.3 g/mol
Melting Pt:162…167 °C
Storage Temperature:Refrigerator
EINECS:216-826-1
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- Specific optical rotation:+26° to +29° (20°C, 589 nm) (c=2, 1N HCl)
Specification Test Results
Frequently Bought Together