

Bis(trimethylsilyl)methane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2117-28-4
:
Formula:C₇H₂₀Si₂
MDL Number:MFCD00008275
:160.405 g/mol
Boiling Pt:132…133 °C
Melting Pt:–71 °C
Density:0.750
Flash Pt:12 °C (53 °F)
Storage Temperature:Ambient
EINECS:218-322-7
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together