1,1'-Carbonyldi(1H-1,2,4-triazole), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Di(1H-1,2,4-triazol-1-yl)methanone
CAS Number:41864-22-6
:
Formula:C₅H₄N₆O
MDL Number:MFCD00043399
:164.1249 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together