HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate), Sigma-Aldrich®
: Merck
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate), Sigma-Aldrich®
: Merck
:94790-37-1
:
:C₁₁H₁₆F₆N₅OP
:MFCD00075445
:379.2 g/mol
Frequently Bought Together
VWR®, Nitrile Gloves
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Tetramethylurea, Sigma-Aldrich®
: Merck