HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: HBTU, 1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)
CAS Number:94790-37-1
:
Formula:C₁₁H₁₆F₆N₅OP
MDL Number:MFCD00075445
:379.2 g/mol
Melting Pt:∼ 200 °C (dec.)
EINECS:423-020-5
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together