
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:125700-67-6
:
Formula:C₁₁H₁₆BF₄N₅O
MDL Number:MFCD00077413
:321.1 g/mol
Melting Pt:195…198 °C (dec.)
EINECS:423-040-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together