Order Entry
Austria
ContactUsLinkComponent
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 99%
Flammable
Irritant
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 99%
Certificates
undefined
Flammable
Irritant
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 99%

Specification Test Results

  • Appearance:
    White to cream Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    ≥200 °C
  • Loss on drying:
    ≤1 %
  • HPLC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together