Isosorbide dimethyl ether, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan
- CAS Number:5306-85-4
- Formula:C₈H₁₄O₄
- MDL Number:MFCD00064828
- Molecular Weight:174.20 g/mol
- :M-Clarity™ quality level = MQ100
- EINECS:226-159-8
2 Options Available Below
Product Details & Documents
Synonyms: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan
- CAS Number:5306-85-4
- Formula:C₈H₁₄O₄
- MDL Number:MFCD00064828
- Molecular Weight:174.20 g/mol
- :M-Clarity™ quality level = MQ100
- EINECS:226-159-8
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



