Isosorbide dimethyl ether ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 5306-85-4 Molecular Formula: C8H14O4 Formula Weight: 174.20 Flash Point: 108°(226°F) Refractive Index: 1.4620 Melting Point: -70° Boiling Point: 93-95°/0.1mm Beilstein Reference: 80854 MDL No.: MFCD00064828
Synonyms: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan
- CAS Number:5306-85-4
- Formula:C₈H₁₄O₄
- MDL Number:MFCD00064828
- Molecular Weight:174.20 g/mol
- Boiling Pt:93…95 °C (0.1 mmHg)
- Melting Pt:–70 °C
- Density:1.165
- Flash Pt:108 °C (226 °F)
- Storage Temperature:Ambient
- EINECS:226-159-8
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 5306-85-4 Molecular Formula: C8H14O4 Formula Weight: 174.20 Flash Point: 108°(226°F) Refractive Index: 1.4620 Melting Point: -70° Boiling Point: 93-95°/0.1mm Beilstein Reference: 80854 MDL No.: MFCD00064828
Synonyms: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan
- CAS Number:5306-85-4
- Formula:C₈H₁₄O₄
- MDL Number:MFCD00064828
- Molecular Weight:174.20 g/mol
- Boiling Pt:93…95 °C (0.1 mmHg)
- Melting Pt:–70 °C
- Density:1.165
- Flash Pt:108 °C (226 °F)
- Storage Temperature:Ambient
- EINECS:226-159-8



