Order Entry
Austria
Orders LinkContactUsLinkComponent
Isosorbide dimethyl ether 99%

Isosorbide dimethyl ether 99%

Isosorbide dimethyl ether 99%
Certificates
Isosorbide dimethyl ether 99%

2 Options Available Below

Product Details & Documents
Synonyms:  (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan
  • CAS Number:5306-85-4
  • Formula:C₈H₁₄O₄
  • MDL Number:MFCD00064828
  • Molecular Weight:174.20 g/mol
  • Boiling Pt:93…95 °C (.1hPa)
  • Flash Pt:120 °C
  • Storage Temperature:Ambient
  • EINECS:226-159-8

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4600 to 1.4620 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 47,70 €
    47,70 €
    1 * 25 g
       47,70 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 133,00 €
    133,00 €
    1 * 100 g
       133,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.