Iminostilbene
Catalog # PROOMM0076.08
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:193.25 g/mol
EINECS:205-970-0
CAS Number:256-96-2
Formula:C₁₄H₁₁N
MDL Number:MFCD00005071