Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Estrone

Glass bottle for solids, 100 mg
Catalog # PROOICRS0302
Estrone
Catalog # PROOICRS0302
CAS Number:  53-16-7
Estrone

Glass bottle for solids, 100 mg

CAS Number:  53-16-7
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
  • CAS Number:53-16-7
  • Formula:C₁₈H₂₂O₂
  • MDL Number:MFCD00003620
  • EINECS:200-164-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications