Estrone
Supplier: Cayman Chemical
Jump to:
About this item
- CAS Number:53-16-7
- Formula:C₁₈H₂₂O₂
- MDL Number:MFCD00003620
- EINECS:200-164-5
4 Options Available Below
Product Details & Documents
- CAS Number:53-16-7
- Formula:C₁₈H₂₂O₂
- MDL Number:MFCD00003620
- EINECS:200-164-5
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



