Order Entry
Austria
ContactUsLinkComponent
Estriol
Estriol
Catalog # MOLE72030665-1G
Supplier:  Molekula
CAS Number:  
Estriol
Catalog # MOLE72030665-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
EINECS:200-022-2
CAS Number:50-27-1
Formula:C₁₈H₂₄O₃
MDL Number:MFCD00003691