Order Entry
Austria
ContactUsLinkComponent
Pesticide reference standard, Chlordane 2000 µg/ml in methanol, Dr Ehrenstorfer
Pesticide reference standard, Chlordane 2000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA11200000ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Chlordane 2000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA11200000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    57-74-9
  • Description:
    Chlordane 2,000 µg/ml in methanol
  • Environmentally Preferable:
  • Formula:
    C₁₀H₆Cl₈
  • Description:
    Chlordane

Specifications

About this item