Order Entry
Austria
ContactUsLinkComponent
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%
Certificates
undefined
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Off-white to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    55 to 59 °C
  • GC:
    ≥97.5 %
  • UV:
    (1 % in CH2Cl2) 1 cm cell

Specification Test Results

Related Information

Frequently Bought Together