3,5-Dimethylpiperidin (mixture of cis- and trans- isomers), Sigma-Aldrich®
Catalog # SIAL186104-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:113.20 g/mol
:M-Clarity™ quality level = MQ100
EINECS:252-730-6
CAS Number:35794-11-7
Formula:C₇H₁₅N
MDL Number:MFCD00005996