Hexaammineruthenium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:15305-72-3
:
Formula:H₁₈Cl₂N₆Ru
MDL Number:MFCD00011477
:274.16 g/mol
:M-Clarity™ quality level = MQ100
EINECS:239-342-2
Frequently Bought Together