
Copper(II) hexafluoroacetylacetonate hydrate ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:155640-85-0
Formula:C₁₀H₂CuF₁₂O₄·xH₂O
MDL Number:MFCD00151019
:477.64 g/mol (anhy)
Boiling Pt:220 °C (dec.)
Melting Pt:∼ 100 °C
Storage Temperature:Ambient
EINECS:238-850-1
:GHS07
Related Information
Frequently Bought Together